Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Unisette™ M406
Blue
Catalog # 720-2959
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Catalog # 720-2959
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Biopsy processing/embedding cassettes, Unisette™ M406
Blue
Price
272,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
  • Moulded from a special high density polymer
  • Resistant to the chemical action of most histological solvents
Specifications
  • Colour:
    Blue
  • Seq:
    16