Microplates, opaque OptiPlates™
OptiiPlate™-384 microplates, 384, White, Case of 50
PERK6007290
These plates are used for general isotopic, fluorescence, LANCE™, [FP]², AlphaScreen™ or luminescence assays.
- Made of polystyrene
- No. of wells:384
- Packed:Case of 50
- Colour:White
- Description:OptiiPlate™-384 microplates
- Seq:04