Order Entry
Export
Orders LinkContactUsLinkComponent
Millicell® EZ Slides

Millicell® EZ Slides

4 well glass chamber slide, well surface area: 1,7 cm², well volume: 500 - 1700 μl, L×W×H: 75×25×1 mm, slide only, 96, Soda-lime glass slide, PP, +
Catalog # MILFPEZGS0496
Supplier:   Merck
Millicell® EZ Slides
Catalog # MILFPEZGS0496
Supplier:  Merck
Millicell® EZ Slides
Millicell® EZ Slides

4 well glass chamber slide, well surface area: 1,7 cm², well volume: 500 - 1700 μl, L×W×H: 75×25×1 mm, slide only, 96, Soda-lime glass slide, PP, +

Price
963,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Millicell® EZ Slide allows the user to grow, fix, and stain cells directly on a microscope slide. There is no need to disassemble the unit or transfer cells from one device to another prior to analysis.

The polypropylene wells are resistant to most fixatives used, so the cells can be grown on the slide and stained right in the well. Once stained, the microscope slide can be easily removed by pressing down the four side tabs of the unit and lifting the wells off of the slide.

This easy slide removal reduces the risk of damaging the cells and no special tools are required to remove the wells from the slide. Raised corner ridges on the cover facilitate stacking of multiple units in the incubator, or they can be incubated in the tray supplied with the units.

Raised corner ridges on the lid facilitate stacking of multiple units.
Techniques: Cell based assay, suitable cell culture, mammalian: suitable immunocytochemistry: suitable.

Specifications
  • No. of wells:
    96
  • Sterile:
    +
  • Description:
    4 well glass chamber slide, well surface area: 1,7 cm², well volume: 500 - 1700 μl, L×W×H: 75×25×1 mm, slide only
  • Material:
    PP
  • Seq:
    02
  • Slide type:
    Soda-lime glass slide
Accessories

Product Information

  • Catalog #
    Material
    Availability
    Price
  • Microscope slide holder for 4 and 8 well units, holds two slides, L×W×H: 128×86×147/17/95 mm, black
    Microscope slide holder for 4 and 8 well units, holds two slides, L×W×H: 128×86×147/17/95 mm, black
    MILFPEZXMSH01
    PS
    Log in for availability
    1 * 1 items - 203,00 €