Order Entry
Export
Orders LinkContactUsLinkComponent
Viscosity standards, general purpose

Viscosity standards, general purpose

S2000, 7300 mm²/s, Glass bottle, 500 mL
Catalog # CANN9727-C55-500ML
Supplier:   CANNON INSTRUMENT
Viscosity standards, general purpose
Catalog # CANN9727-C55-500ML
Supplier:  CANNON INSTRUMENT
CAS Number:  
Viscosity standards, general purpose

S2000, 7300 mm²/s, Glass bottle, 500 mL

CAS Number:  
Price
93,49 €
Log in for availability
1 * 500 mL
Product Details & Documents

These certified standards are useful for calibration and verification procedures.

  • It is suitable for all types of viscometers
  • Viscosity data quoted at 20 °C
  • It also helps ensures the reliability of measurements

Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • No.:
    S2000
  • Viscosity:
    7300 mm²/s

Specifications