Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
uffer pH 7,00 (yellow) +/-0,01 @25C (Bag In Box) 1 * 5 L
Catalog # REGEBPH09
Supplier:
REAGECON
Expand image
uffer pH 7,00 (yellow) +/-0,01 @25C (Bag In Box) 1 * 5 L
Catalog # REGEBPH09
Supplier:
REAGECON