Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COVER GLASS 5MM DIA. NO.1 0.13-0.16MM 1 * 1.000 items
Catalog # MARI0111990395
Supplier:
MARIENFELD
Expand image
COVER GLASS 5MM DIA. NO.1 0.13-0.16MM 1 * 1.000 items
Catalog # MARI0111990395
Supplier:
MARIENFELD