Accessories for PURELAB Flex Systems
UV lamp 185/254 nm for the reduction of TOC and bacteria.
For PURELAB® flex 2, 3 and 4.
For PURELAB® flex 2, 3 and 4.
Catalog # ELGALC210-02
Supplier: ELGA LabWater
Jump to:
Product Details & Documents
Specifications
- Description:UV lamp 185/254 nm for the reduction of TOC and bacteria.
For PURELAB® flex 2, 3 and 4.