Order Entry
Export
Orders LinkContactUsLinkComponent
Cover glasses

Cover glasses

Cover glass, 0,13 - 0,16 mm, 5 mm, Float glass
Catalog # BELC1943-00005
Supplier:   BELLCO GLASS
Cover glasses
Catalog # BELC1943-00005
Supplier:  BELLCO GLASS
Cover glasses
Cover glasses

Cover glass, 0,13 - 0,16 mm, 5 mm, Float glass

Price
71,10 €
Log in for availability
1 * 100 items
Product Details & Documents

Round cover slips made of glass.

Specifications
  • Ø:
    5 mm
  • Description:
    Cover glass
  • Material:
    Float glass
  • Thickness:
    0,13 - 0,16 mm