Compact burette, BLAUBRAND®, cl.AS, DE-M, 25:0,05ml, Boro 3.3,PTFE stopcock, Schellbach , with individual USP certificate 1 * 1 items
Catalog # BRNDUSP13916
Compact burette, BLAUBRAND®, cl.AS, DE-M, 25:0,05ml, Boro 3.3,PTFE stopcock, Schellbach , with individual USP certificate 1 * 1 items
Catalog # BRNDUSP13916