Orders LinkContactUsLinkComponent
Disposable syringes
Disposable syringes
Luer lock syringe, 3 ml
Catalog # NSCAS7515-3
Disposable syringes
Catalog # NSCAS7515-3
Disposable syringes
Luer lock syringe, 3 ml
Price
78,60 €
Log in for availability
1 * 100 items
Product Details & Documents
These syringes are an ideal choice for all syringe filter applications. They feature a two-part construction using PP barrels and PE plungers. This eliminates the need for rubber or synthetic plunger gaskets and requires no silicone or oil lubricant in the barrel.
  • Non sterile
  • Bulk packed
Specifications
  • Volume:
    3 ml
  • Description:
    Luer lock syringe
  • Seq:
    01