Multi-element noble metals standards, VHG
Noble metals standard: Au; Ir; Os; Pd; Pt; Re; Rh; Ru, 100 mg/L, 20% HCl, 500 mL
Catalog # PROOVHG-SM40-500
Supplier: LGC Standards PROMOCHEM
Multi-element noble metals standards, VHG
Catalog # PROOVHG-SM40-500
Supplier: LGC Standards PROMOCHEM
CAS Number:
Jump to:
Product Details & Documents
Multi element standards for Au, Ir, Os, Pd, Pt, Re, Rh, Ru at 100 µg/ml in 20% HCl.
Specifications
Specifications
- Concentration:100 mg/L
- Matrix:20% HCl
- Pk:500 mL
- Description:Noble metals standard: Au; Ir; Os; Pd; Pt; Re; Rh; Ru



