Order Entry
Export
Orders LinkContactUsLinkComponent
 

Multi-element noble metals standards, VHG

Noble metals standard: Au; Ir; Os; Pd; Pt; Re; Rh; Ru, 100 mg/L, 20% HCl, 100 mL
  PROOVHG-SM40-100
Multi-element noble metals standards, VHG
  PROOVHG-SM40-100
 :  
Multi-element noble metals standards, VHG

Noble metals standard: Au; Ir; Os; Pd; Pt; Re; Rh; Ru, 100 mg/L, 20% HCl, 100 mL

 :  
 
283,00 €
   
1 * 100 mL
 

Multi element standards for Au, Ir, Os, Pd, Pt, Re, Rh, Ru at 100 µg/ml in 20% HCl.

 

 

  • Concentration:
    100 mg/L
  • Matrix:
    20% HCl
  • Pk:
    100 mL
  • Description:
    Noble metals standard: Au; Ir; Os; Pd; Pt; Re; Rh; Ru