Organic reference standards, fluvoxamines, Ph. Eur.
Fluvoxamine impurity D, 5-methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one, Ph. Eur. standard, Reference standard, 10 mg
Catalog # EDQMY0000916
Supplier: EDQM
Organic reference standards, fluvoxamines, Ph. Eur.
Catalog # EDQMY0000916
Supplier: EDQM
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:10 mg
- Standards:Reference standard
- Description:Fluvoxamine impurity D, 5-methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one, Ph. Eur. standard
- Cat. No.:EDQMY0000916
- Formula:C₁₃H₁₅F₃O₂
- Description:Fluvoxamine impurity D, 5-methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one
- CAS No.:61718-80-7



