ContactUsLinkComponent
Rotavapor R-300 B-305, no joint., A, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: Diagonal (A) 1 * 1 items
Catalog # BUCH11R300150A001
Supplier:  BUCHI
Rotavapor R-300 B-305, no joint., A, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: Diagonal (A) 1 * 1 items
Catalog # BUCH11R300150A001
Supplier:  BUCHI