ContactUsLinkComponent
Rotavapor R-300 B-301, SJ24/40, A, WB, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 24/40 Glass set: Diagonal (A) int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300112A011
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ24/40, A, WB, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 24/40 Glass set: Diagonal (A) int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300112A011
Supplier:  BUCHI