ContactUsLinkComponent
Rotavapor R-300 no bath, SJ29/32, V, WB, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: Vertical (V) int.: Woulf f b., w/o int. 1 * 1 items
Catalog # BUCH11R300101V011
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ29/32, V, WB, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: Vertical (V) int.: Woulf f b., w/o int. 1 * 1 items
Catalog # BUCH11R300101V011
Supplier:  BUCHI