ContactUsLinkComponent
Rotavapor R-300 no bath, no joint., C, I-300P, 230V. . Lift System: Elec. Heating Bath: w/o Glass set: Cold trap, int.: I-300 Pro, W oulff bottle 1 * 1 items
Catalog # BUCH11R300200C031
Supplier:  BUCHI
Rotavapor R-300 no bath, no joint., C, I-300P, 230V. . Lift System: Elec. Heating Bath: w/o Glass set: Cold trap, int.: I-300 Pro, W oulff bottle 1 * 1 items
Catalog # BUCH11R300200C031
Supplier:  BUCHI