ContactUsLinkComponent
Rotavapor R-300 B-305, no joint., V, P+G, I-300, 230V. . Elec. lift, B-305 bath, Glass set: Vertical (V) P+G coating, int.: I-300, W oulff bottle 1 * 1 items
Catalog # BUCH11R300250V121
Supplier:  BUCHI
Rotavapor R-300 B-305, no joint., V, P+G, I-300, 230V. . Elec. lift, B-305 bath, Glass set: Vertical (V) P+G coating, int.: I-300, W oulff bottle 1 * 1 items
Catalog # BUCH11R300250V121
Supplier:  BUCHI