Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Rotavapor R-300 B-305, SJ29/32, V, I-300, VU, 230V. . Elec. lift, B-305 bath, SJ 29/32 Glass set: Vertical (V) int.: I-300, valve u nit 1 * 1 items
BUCH11R300251V041
:
BUCHI
Rotavapor R-300 B-305, SJ29/32, V, I-300, VU, 230V. . Elec. lift, B-305 bath, SJ 29/32 Glass set: Vertical (V) int.: I-300, valve u nit 1 * 1 items
BUCH11R300251V041
:
BUCHI