Rotavapor R-300 B-305, SJ24/40, A, P+G, I-300, VU, 230V. . Elec. lift, B-305 bath, SJ 24/40 Glass set: Diagonal (A) P+G coating, in t.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300252A141
Rotavapor R-300 B-305, SJ24/40, A, P+G, I-300, VU, 230V. . Elec. lift, B-305 bath, SJ 24/40 Glass set: Diagonal (A) P+G coating, in t.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300252A141