Orders LinkContactUsLinkComponent
Test strips, peroxide, Dosatest
Test strips, peroxide, Dosatest
Peroxide test strips 0 - 100 mg/l, 0 · 1 · 3 · 10 · 30 · 100, 100
Catalog # 85434.601
Supplier:   VWR Chemicals
Test strips, peroxide, Dosatest
Catalog # 85434.601
Supplier:  VWR Chemicals
CAS Number:  
Test strips, peroxide, Dosatest
Test strips, peroxide, Dosatest
Peroxide test strips 0 - 100 mg/l, 0 · 1 · 3 · 10 · 30 · 100, 100
CAS Number:  
Price
29,20 €
Log in for availability
1 * 100 items
Product Details & Documents
A ready to use kit that fits into your lab coat pocket. Intended for water-based samples
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor your desinfection processes. Semi-quantitative results can be obtained between 10 and 120 seconds.
The colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6×95 mm.
Specifications
Specifications
  • Pk:
    100
  • Description:
    Peroxide test strips 0 - 100 mg/l
  • Range:
    0 · 1 · 3 · 10 · 30 · 100
Specifications