Test strips, peroxide, Dosatest
Peroxide test strips 0 - 25 mg/l, 0 · 0,5 · 2 · 5 · 10 · 25, 100
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
A ready to use kit that fits into your lab coat pocket. Intended for water-based samples
- Rapid and convenient
- Easy to handle
- Choice of ranges for required precision
Ideal to monitor your desinfection processes. Semi-quantitative results can be obtained between 10 and 120 seconds.
The colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6×95 mm.
Documents
Specifications
Specifications
- Pk:100
- Description:Peroxide test strips 0 - 25 mg/l
- Range:0 · 0,5 · 2 · 5 · 10 · 25