Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Dosatest, pH indicator test strips, non bleeding

VWR® Dosatest, pH indicator test strips, non bleeding

4,0 - 7,0, 4,0 · 4,4 · 4,7 · 5,0 · 5,3 · 5,5 · 5,8 · 6,1 · 6,5 · 7,0
Catalog # 85423.601
Supplier:   VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Catalog # 85423.601
Supplier:  VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
VWR® Dosatest, pH indicator test strips, non bleeding

4,0 - 7,0, 4,0 · 4,4 · 4,7 · 5,0 · 5,3 · 5,5 · 5,8 · 6,1 · 6,5 · 7,0

Price
13,90 €
Log in for availability
1 * 100 items
Product Details & Documents

Dosatest test strips provide precise and rapid pH determination, with up to four different colour blocks for each pH value.

  • Easier to work with due to the long handle
  • Colour-fixed reactive pads
  • Brilliant colour chart

pH indicator is chemically bound onto the cellulose fibre preventing bleeding. This effectively protects the sample against contamination and enables measurements to be taken even in weak buffered or strong alkaline solutions.

Better access to different types of containers preventing fingers coming into contact with the sample.

Delivery information: Supplied in a box with 100 test strips 6×85 mm.

Specifications
  • pH gradation:
    4,0 · 4,4 · 4,7 · 5,0 · 5,3 · 5,5 · 5,8 · 6,1 · 6,5 · 7,0
  • pH range:
    4,0 - 7,0
  • Seq:
    28