Methyl blue
25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 28983-56-4 Molecular Formula: C37H27N3Na2O9S3 Formula Weight: 799.80 Physical Form: Powder Appearance: Blue Odour: Odourless MDL No.: MFCD00058509
Synonyms: Aniline blue sodium salt, [[4-[bis[4-[(sulphophenyl)amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]amino]-benzenesulphonic acid sodium salt (1:2), Poirriers blue, Cotton blue, Water blue, C.I. 42780, Aniline blue, water soluble, Disodium [[4-[bis[4-[(sulphonatophenyl)amino]phenyl]methylene]cyclohexa-2,5-dien-1-ylidene]amino]benzenesulphonate, Acid Blue 93
- CAS Number:28983-56-4
- Formula:C₃₇H₂₇N₃Na₂O₉S₃
- MDL Number:MFCD00058509
- Molecular Weight:799.81 g/mol
- Storage Temperature:Ambient
- EINECS:249-352-9
Documents
Specifications
Specifications
- Pk:25 g



