Mobile storage cabinets
Mobile storage cabinet, With front open, 900×500×590 mm
KOTT315-00128
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
- Description:Mobile storage cabinet
- Seq:22
- W×D×H:900×500×590 mm
- Doors:With front open