Order Entry
Export
Orders LinkContactUsLinkComponent
 

Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height, 1 door right, 450×516×740 mm
Catalog # KOTT315-00124
Supplier:   KOETTERMANN
Mobile storage cabinets
Catalog # KOTT315-00124
Supplier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height, 1 door right, 450×516×740 mm

Price
627,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Epoxy powder coated steel cabinets

  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifications
  • Description:
    Mobile storage cabinet, with 1 drawer of 150 mm height
  • Seq:
    16
  • W×D×H:
    450×516×740 mm
  • Doors:
    1 door right