Mobile storage cabinets
Mobile storage cabinet with 1 shelf, With front open, 450×500×740 mm
KOTT315-00116
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
- Description:Mobile storage cabinet with 1 shelf
- Seq:10
- W×D×H:450×500×740 mm
- Doors:With front open