Mobile storage cabinets
Mobile storage cabinet with 1 shelf, lockable, 1 door right, 450×516×590 mm
Catalog # KOTT315-00113
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifications
- Description:Mobile storage cabinet with 1 shelf, lockable
- Seq:07
- W×D×H:450×516×590 mm
- Doors:1 door right