Orders LinkContactUsLinkComponent
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet with 1 shelf, lockable, 1 door left, 450×516×590 mm
Catalog # KOTT315-00111
Supplier:   KOETTERMANN
Mobile storage cabinets
Catalog # KOTT315-00111
Supplier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet with 1 shelf, lockable, 1 door left, 450×516×590 mm
Price
472,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifications
  • Description:
    Mobile storage cabinet with 1 shelf, lockable
  • Seq:
    05
  • W×D×H:
    450×516×590 mm
  • Doors:
    1 door left