Order Entry
Export
Orders LinkContactUsLinkComponent
 

Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height, 2 doors, 1200×516×740 mm
  KOTT315-00102
Mobile storage cabinets
  KOTT315-00102
Mobile storage cabinets
Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height, 2 doors, 1200×516×740 mm

 
962,00 €
   
1 * 1 items
 

Epoxy powder coated steel cabinets

  • Optional locks
  • Fitted with castors
  • Removable shelves
 
  • Description:
    Mobile storage cabinet, with 1 drawer of 150 mm height
  • Seq:
    47
  • W×D×H:
    1200×516×740 mm
  • Doors:
    2 doors