Mobile storage cabinets
Mobile storage cabinet, lockable, 2 doors, 1200×516×740 mm
Catalog # KOTT315-00101
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifications
- Description:Mobile storage cabinet, lockable
- Seq:46
- W×D×H:1200×516×740 mm
- Doors:2 doors