Orders LinkContactUsLinkComponent
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, 2 doors, 1200×516×740 mm
Catalog # KOTT315-00100
Supplier:   KOETTERMANN
Mobile storage cabinets
Catalog # KOTT315-00100
Supplier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, 2 doors, 1200×516×740 mm
Price
821,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifications
  • Description:
    Mobile storage cabinet
  • Seq:
    45
  • W×D×H:
    1200×516×740 mm
  • Doors:
    2 doors