Order Entry
Export
Orders LinkContactUsLinkComponent
Mobile storage cabinets

Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height and 1 drawer of 300 mm height, lockable, 2 drawers, 1200×516×590 mm
Catalog # KOTT315-00093
Supplier:   KOETTERMANN
Mobile storage cabinets
Catalog # KOTT315-00093
Supplier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height and 1 drawer of 300 mm height, lockable, 2 drawers, 1200×516×590 mm

Price
1 052,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Epoxy powder coated steel cabinets

  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifications
  • Description:
    Mobile storage cabinet, with 1 drawer of 150 mm height and 1 drawer of 300 mm height, lockable
  • Seq:
    38
  • W×D×H:
    1200×516×590 mm
  • Doors:
    2 drawers