Mobile storage cabinets
Mobile storage cabinet with 1 shelf, With front open, 1200×500×590 mm
Catalog # KOTT315-00092
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifications
- Description:Mobile storage cabinet with 1 shelf
- Seq:37
- W×D×H:1200×500×590 mm
- Doors:With front open