NMR sample tube washer
NMR sample tube washer, acc. to Swiegot, complete, to fit bottles with socket ST 29/32
Catalog # WITG2.900.029
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Complete with 2 PTFE-adapters to fir 5 cuvets or NMR tubes of 5 and 10 mm diameter in different lengths. Vacuum operated washers allowing the individual washing of 1 to 5 tube, complete with caps and FEPcappilaries (box 1 pce.).
Specifications
- Description:NMR sample tube washer, acc. to Swiegot, complete, to fit bottles with socket ST 29/32
- Seq:01