Order Entry
Export
Orders LinkContactUsLinkComponent
Ghost Dye™ Red 710

Ghost Dye™ Red 710

Red 710, 500 Tests
Catalog # 13-0871-T500
Supplier:   Tonbo Biosciences
Ghost Dye™ Red 710
Catalog # 13-0871-T500
Supplier:  Tonbo Biosciences
CAS Number:  
Ghost Dye™ Red 710
Ghost Dye™ Red 710

Red 710, 500 Tests

CAS Number:  
Price
193,00 €
Log in for availability
1 * 500 Tests
Product Details & Documents

Ghost Dye Red 710 is an amine reactive viability dye, that can be used to discriminate viable from non-viable mammalian cells in flow cytometry applications.

This dye irreversibly binds free amines available on the cell surface, as well as intracellular free amines exposed in cells with compromised cell membranes.

Necrotic cells with compromised membranes will react with significantly more Ghost Dye Red 710 dye than viable cells in the same sample and therefore exhibit much greater fluorescence intensity, allowing exclusion of these cells from analysis.

Specifications

Specifications

  • Pk:
    500 Tests
  • Description:
    Red 710
  • Cat. No.:
    13-0871-T500
  • Storage temperature:
    At ‒20 °C protected from light and moisture
  • Application:
    Flow cytometry

Specifications