Order Entry
Export
Orders LinkContactUsLinkComponent
Inlet liners

Inlet liners

Split/splitless, with wool, universal for most common analyses, 5,0, 3,5 mm, 95 mm
Catalog # REST20956-5
Inlet liners
Catalog # REST20956-5
Supplier:  
Inlet liners
Inlet liners

Split/splitless, with wool, universal for most common analyses, 5,0, 3,5 mm, 95 mm

Discontinued
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    5,0
  • Ø int.:
    3,5 mm
  • Description:
    Split/splitless, with wool, universal for most common analyses
  • Length:
    95 mm
  • Seq:
    075