Order Entry
Export
Orders LinkContactUsLinkComponent
 

Inlet liners

Splitless, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSL, 8,0 mm, 3,0 mm, 105 mm
Catalog # REST20943
Supplier:   Restek
Inlet liners
Catalog # REST20943
Supplier:  Restek
Inlet liners
Inlet liners

Splitless, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSL, 8,0 mm, 3,0 mm, 105 mm

Price
249,00 €
Log in for availability
1 * 5 items
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    8,0 mm
  • Ø int.:
    3,0 mm
  • Description:
    Splitless, for trace samples, for Trace™, 8000, 8000 TOP & Focus SSL
  • Length:
    105 mm
  • Seq:
    105