Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) sulphide 90%, pure

Lead(II) sulphide 90%, pure

Plastic bottle, 1 kg
Catalog # ACRO453970010
Irritant
Health
Environment
Lead(II) sulphide 90%, pure
Catalog # ACRO453970010
CAS Number:  1314-87-0
Lead(II) sulphide 90%, pure
Lead(II) sulphide 90%, pure

Plastic bottle, 1 kg

CAS Number:  1314-87-0
Price
99,40 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Lead sulphide
  • CAS Number:1314-87-0
  • Formula:̍PbS
  • MDL Number:MFCD00016280
  • Molecular Weight:239.27 g/mol
  • Boiling Pt:1281.1 °C
  • Melting Pt:1113.9 °C
  • Density:7,5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:3077
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Grey to purple Powder
  • Assay:
    ≥78 % (Pb)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications