Propidium Iodide 95%
Glass bottle, 1 g
Supplier: Thermo Fisher Scientific
Propidium Iodide 95%
Catalog # ACRO440300010
Supplier: Thermo Fisher Scientific
CAS Number: 25535-16-4
Jump to:
Product Details & Documents
Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
- CAS Number:25535-16-4
- Formula:C₂₇H₃₄I₂N₄
- MDL Number:MFCD00011921
- Molecular Weight:668.4 g/mol
- Melting Pt:220…225 °C (dec.)
- Storage Temperature:Refrigerator
Documents
ACRO44030
SDS
Specifications
Specification Test Results
- Appearance:Red Powder
- Infrared spectrum:Conforms
- Melting point:200 to 225 °C
- Titration Argentometric:≥94 %
- Water:≤5 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



