Order Entry
Export
Orders LinkContactUsLinkComponent
 

Crystal violet ACS

Glass bottle, 250 g
Catalog # ACRO447572500
Irritant
Health
Corrosive
Environment
Crystal violet ACS
Catalog # ACRO447572500
CAS Number:  548-62-9
Crystal violet ACS
Crystal violet ACS

Glass bottle, 250 g

CAS Number:  548-62-9
Price
201,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
  • CAS Number:548-62-9
  • Formula:C₂₅H₃₀ClN₃
  • MDL Number:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Melting Pt:173 °C (dec.)
  • Density:1,19 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:3077
  • ADR:9,III

 

Specifications

Specification Test Results

  • Appearance:
    Dark green Powder or crystals
  • Assay:
    ≥90.0 % (on dry substance)
  • Loss on drying:
    ≤7.5 %
  • Absorbance characteristics:
    (0.98-1.9) Passes test
  • Sensitivity:
    Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g

Specifications