Sodium hydroxide, pellets, pure
Fibre drum, 25 kg
Supplier: Thermo Fisher Scientific
Sodium hydroxide, pellets, pure
Catalog # ACRO447280250
Supplier: Thermo Fisher Scientific
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:1390 °C (760 mmHg)
- Melting Pt:318 °C
- Density:2,13 g/cm³ (20 °C)
- Storage Temperature:Ambient
- UN:1823
- ADR:8,II
Specifications
Specification Test Results
- Appearance:White to off-white Pellets
- Titration with HCl:≥97.5 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO4):≤0.005 %
- Sodium carbonate (Na2CO3):≤2.0 %
- Iron (Fe):≤0.001 %
- Heavy metals (as Pb):≤20 ppm
Specifications
- Pack type:Fibre drum
- Pk:25 kg



