Order Entry
Export
Orders LinkContactUsLinkComponent
 

Nitromethane 96% for HPLC

Glass bottle, 100 mL
Catalog # ACRO445671000
Flammable
Irritant
Health
Nitromethane 96% for HPLC
Catalog # ACRO445671000
CAS Number:  75-52-5
Nitromethane 96% for HPLC
Nitromethane 96% for HPLC

Glass bottle, 100 mL

CAS Number:  75-52-5
Price
29,10 €
Log in for availability
1 * 100 mL
Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Boiling Pt:101.2 °C (1013 hPa)
  • Melting Pt:–29 °C
  • Density:1.12 g/cm³
  • Flash Pt:35 °C
  • Storage Temperature:Ambient
  • UN:1261
  • ADR:3,II

 

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.03 %
  • GC:
    ≥95 %
  • UV:
    at 380 nm A: ≤1
  • Color scale:
    ≤10 APHA

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications