5,7-Dichloro-1,6-naphthyridine 97%
Glass bottle, 1 g
Supplier: Thermo Fisher Scientific
5,7-Dichloro-1,6-naphthyridine 97%
Catalog # ACRO440670010
Supplier: Thermo Fisher Scientific
CAS Number: 337958-60-8
Jump to:
Product Details & Documents
- CAS Number:337958-60-8
- Formula:C₈H₄Cl₂N₂
- MDL Number:MFCD11518965
- Molecular Weight:199.0368 g/mol
- Melting Pt:138…140 °C
- Storage Temperature:Ambient
Documents
ACRO44067
SDS
Specifications
Specification Test Results
- Appearance:White to yellow to light orange Crystalline powder or solid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



