Thymol blue ACS
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-61-9
- Formula:C₂₇H₃₀O₅S
- MDL Number:MFCD00005869
- Molecular Weight:466.6 g/mol
- Merck Index:12,09541
- Melting Pt:221 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-973-3
Documents
ACRO42093
SDS
Specifications
Specification Test Results
- Appearance:Dark green to dark brown Crystalline powder
- Infrared spectrum:Conforms
- Visual transition interval:from pH 1.2 (pink) to pH 2.8 (yellow)
- Clarity of solution:passes test
Specifications
- Pack type:Glass bottle
- Pk:100 g



