Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) nitrate ACS

Lead(II) nitrate ACS

Plastic bottle, 2,5 kg
Catalog # ACRO423850025
Irritant
Health
Environment
Oxidizing
Corrosive
Lead(II) nitrate ACS
Catalog # ACRO423850025
CAS Number:  10099-74-8
Lead(II) nitrate ACS
Lead(II) nitrate ACS

Plastic bottle, 2,5 kg

CAS Number:  10099-74-8
Price
255,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Lead dinitrate
  • CAS Number:10099-74-8
  • Formula:Pb(NO₃)₂
  • MDL Number:MFCD00011153
  • Molecular Weight:331.21 g/mol
  • Merck Index:13,05432
  • Melting Pt:470 °C (dec.)
  • Density:4.53 g/cm³
  • Storage Temperature:Ambient
  • EINECS:233-245-9
  • UN:1469
  • ADR:5.1,II

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Titration Complexometric:
    ≥99.0 %
  • Chloride (Cl):
    ≤10 ppm
  • Calcium (Ca):
    ≤50 ppm
  • Copper (Cu):
    ≤20 ppm
  • Iron (Fe):
    ≤10 ppm
  • Potassium (K):
    ≤50 ppm
  • Sodium (Na):
    ≤200 ppm
  • Insoluble matter:
    ≤0.005 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

Specifications