Triethylamine 99.5% for analysis
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Triethylamine 99.5% for analysis
Catalog # 432280025.
Supplier: Thermo Fisher Scientific
CAS Number: 121-44-8
Jump to:
Product Details & Documents
Synonyms: (Diethylamino)ethane, TEA
- CAS Number:121-44-8
- Formula:(C₂H₅)₃N
- MDL Number:MFCD00009051
- Molecular Weight:101.19 g/mol
- Boiling Pt:90 °C (760 mmHg)
- Melting Pt:–115 °C
- Density:0.728 g/cm³
- Flash Pt:–11 °C
- Storage Temperature:Ambient
- UN:1296
- ADR:3,II
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3990 to 1.4010 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥99.4 %
- Color scale:≤15 APHA
- Residue after evaporation:≤0.002 %
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤10 ppm
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Manganese (Mn):≤0.02 ppm
- Nickel (Ni):≤0.02 ppm
- Zinc (Zn):≤0.1 ppm
- Heavy metals (as Pb):≤1 ppm
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



