Order Entry
Export
Orders LinkContactUsLinkComponent
 

Triethylamine 99.5% for analysis

Glass bottle, 1 L
Catalog # 432280010.
Toxic
Corrosive
Flammable
Triethylamine 99.5% for analysis
Catalog # 432280010.
CAS Number:  121-44-8
Triethylamine 99.5% for analysis
Triethylamine 99.5% for analysis

Glass bottle, 1 L

CAS Number:  121-44-8
Price
70,40 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  (Diethylamino)ethane, TEA
  • CAS Number:121-44-8
  • Formula:(C₂H₅)₃N
  • MDL Number:MFCD00009051
  • Molecular Weight:101.19 g/mol
  • Boiling Pt:90 °C (760 mmHg)
  • Melting Pt:–115 °C
  • Density:0.728 g/cm³
  • Flash Pt:–11 °C
  • Storage Temperature:Ambient
  • UN:1296
  • ADR:3,II
Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3990 to 1.4010 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • GC:
    ≥99.4 %
  • Color scale:
    ≤15 APHA
  • Residue after evaporation:
    ≤0.002 %
  • Chloride (Cl):
    ≤10 ppm
  • Sulfate (SO4):
    ≤10 ppm
  • Cadmium (Cd):
    ≤0.05 ppm
  • Calcium (Ca):
    ≤0.5 ppm
  • Chromium (Cr):
    ≤0.02 ppm
  • Cobalt (Co):
    ≤0.02 ppm
  • Copper (Cu):
    ≤0.02 ppm
  • Iron (Fe):
    ≤0.1 ppm
  • Lead (Pb):
    ≤0.1 ppm
  • Magnesium (Mg):
    ≤0.1 ppm
  • Manganese (Mn):
    ≤0.02 ppm
  • Nickel (Ni):
    ≤0.02 ppm
  • Zinc (Zn):
    ≤0.1 ppm
  • Heavy metals (as Pb):
    ≤1 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications