Nitrobenzene 99+% ACS
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:98-95-3
- Formula:C₆H₅NO₂
- MDL Number:MFCD00007043
- Molecular Weight:123.11 g/mol
- Merck Index:13,06621
- Boiling Pt:210.8 °C (1013 hPa)
- Melting Pt:5…6 °C
- Density:1.205 g/cm³
- Flash Pt:88 °C
- Storage Temperature:Ambient
- EINECS:202-716-0
- UN:1662
- ADR:6.1,II
Documents
Specifications
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5500 to 1.5540 (20°C, 589 nm)
- GC:≥99.0 %
- Residue after evaporation:≤0.005 %
- Chloride (Cl):≤5 ppm
- Water soluble titr. acid:≤0.0005 meq/g
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



