Order Entry
Export
Orders LinkContactUsLinkComponent
1,3,2-Dioxathiolane-2,2-dioxide 96%

1,3,2-Dioxathiolane-2,2-dioxide 96%

Glass bottle, 1 g
Catalog # ACRO426630010
Irritant
1,3,2-Dioxathiolane-2,2-dioxide 96%
Catalog # ACRO426630010
CAS Number:  1072-53-3
1,3,2-Dioxathiolane-2,2-dioxide 96%
1,3,2-Dioxathiolane-2,2-dioxide 96%

Glass bottle, 1 g

CAS Number:  1072-53-3
Price
30,10 €
Log in for availability
1 * 1 g
Product Details & Documents

Appearance: White to beige Powder

Synonyms:  1,3,2-Dioxathiolane 2,2-dioxide, Ethylene Sulphate
  • CAS Number:1072-53-3
  • Formula:C₂H₄O₄S
  • MDL Number:MFCD00221769
  • Molecular Weight:124.11 g/mol
  • Melting Pt:95…97 °C
  • Storage Temperature:Refrigerator

Documents

Specifications

Specification Test Results

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    95 to 99 °C
  • GC:
    ≥95.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications